ENA358813269 Enamine

1-(furan-2-yl)-2-methoxyethan-1-amine

  • MDL number MFCD09729980
  • Cas Number : 186029-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97641
Availability details for
Quantity
  • Availability
  • Add to Cart