ENA358813273 Enamine

(2-aminoethyl)(ethyl)propylamine

  • MDL number MFCD01317551
  • Cas Number : 57190-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97647

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-97647
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart