ENA358813283 Enamine

N-(4-hydroxycyclohexyl)-2-(trimethyl-1H-pyrazol-4-yl)acetamide

  • MDL number MFCD12639021
  • Cas Number : 1157107-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-97710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97710
Availability details for
Quantity
  • Availability
  • Add to Cart