ENA358813286 Enamine

1-iodo-3-methanesulfonylpropane

  • MDL number MFCD20414742
  • Cas Number : 1339388-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97718

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97718
Availability details for
Quantity
  • Availability
  • Add to Cart