ENA358813287 Enamine

1-(1-methyl-1H-imidazole-2-carbonyl)isoquinoline

  • MDL number MFCD19555360
  • Cas Number : 1375474-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97719
Availability details for
Quantity
  • Availability
  • Add to Cart