ENA358813292 Enamine

(1-benzofuran-2-ylmethyl)(methyl)amine

  • MDL number MFCD08361685
  • Cas Number : 74377-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97727

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97727
Availability details for
Quantity
  • Availability
  • Add to Cart