ENA358813297 Enamine

2-amino-2-(2-methylphenyl)ethan-1-ol

  • MDL number MFCD11213642
  • Cas Number : 1094627-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97747
Availability details for
Quantity
  • Availability
  • Add to Cart