ENA358813302 Enamine

1-(2,2,2-trifluoroethyl)piperidin-4-ol

  • MDL number MFCD12147930
  • Cas Number : 90633-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97801
Availability details for
Quantity
  • Availability
  • Add to Cart