ENA358813313 Enamine

methyl 2-amino-4-tert-butyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD02082348
  • Cas Number : 478258-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97840
Availability details for
Quantity
  • Availability
  • Add to Cart