ENA358813333 Enamine

3-fluoro-4-(pyridin-2-ylmethoxy)aniline

  • MDL number MFCD08699624
  • Cas Number : 179687-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97894

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97894
Availability details for
Quantity
  • Availability
  • Add to Cart