ENA358813338 Enamine

N-[(4-methoxyphenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD02059792
  • Cas Number : 384858-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97903

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97903
Availability details for
Quantity
  • Availability
  • Add to Cart