ENA358813341 Enamine

3-methoxy-6-methylpyridin-2-amine

  • MDL number MFCD09878490
  • Cas Number : 478913-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97912
Availability details for
Quantity
  • Availability
  • Add to Cart