ENA358813344 Enamine

4-methyl-3,4-dihydro-2H-1,4-benzoxazin-6-amine

  • MDL number MFCD11557839
  • Cas Number : 226571-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97926
Availability details for
Quantity
  • Availability
  • Add to Cart