ENA358813354 Enamine

1-(2,5-difluorophenyl)-3,3,3-trifluoropropan-1-one

  • MDL number MFCD11105455
  • Cas Number : 1152684-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97986
Availability details for
Quantity
  • Availability
  • Add to Cart