ENA358813366 Enamine

2-chloro-4,5-difluoroaniline

  • MDL number MFCD08448325
  • Cas Number : 2613-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-58
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98025

Safety Information

signalword Warning
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P314,P330,P332+P313,P337+P313,P362,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-98025
pictograms GHS07,GHS08
hstatements H302,H312,H315,H319,H332,H373
Availability details for
Quantity
  • Availability
  • Add to Cart