ENA358813368 Enamine

1-ethenyl-4-ethylbenzene

  • MDL number MFCD00085258
  • Cas Number : 3454-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98031
Availability details for
Quantity
  • Availability
  • Add to Cart