ENA358813369 Enamine

3-(aminomethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one hydrochloride

  • MDL number MFCD06738798
  • Cas Number : 367250-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268...270
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98038

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-98038
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart