ENA358813373 Enamine

6-(1-methyl-1H-pyrazol-4-yl)pyridin-2-amine

  • MDL number MFCD12824159
  • Cas Number : 1183196-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98076
Availability details for
Quantity
  • Availability
  • Add to Cart