ENA358813374 Enamine

2-chloro-6-(2,4-dichlorophenoxy)benzoic acid

  • MDL number MFCD12617911
  • Cas Number : 1041555-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98087
Availability details for
Quantity
  • Availability
  • Add to Cart