ENA358813382 Enamine

1-cyclobutylethan-1-ol

  • MDL number MFCD00061039
  • Cas Number : 7515-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98181
Availability details for
Quantity
  • Availability
  • Add to Cart