ENA358813387 Enamine

2-[3-(trifluoromethyl)phenyl]morpholine

  • MDL number MFCD00864259
  • Cas Number : 30914-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98201
Availability details for
Quantity
  • Availability
  • Add to Cart