ENA358813390 Enamine

6-chloro-7-fluoro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11035966
  • Cas Number : 942493-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98208

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98208
Availability details for
Quantity
  • Availability
  • Add to Cart