ENA358813392 Enamine

2,7-dimethyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD15730334
  • Cas Number : 114603-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98210
Availability details for
Quantity
  • Availability
  • Add to Cart