ENA358813395 Enamine

methyl 4-{[4-(hydroxymethyl)piperidin-1-yl]methyl}benzoate

  • MDL number MFCD12622259
  • Cas Number : 1094514-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98330

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98330
Availability details for
Quantity
  • Availability
  • Add to Cart