ENA358813397 Enamine

N-(3,5-dimethoxyphenyl)piperidin-4-amine

  • MDL number MFCD08059696
  • Cas Number : 893763-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98340
Availability details for
Quantity
  • Availability
  • Add to Cart