ENA358813398 Enamine

1-[(3,5-dimethoxyphenyl)methyl]piperazine

  • MDL number MFCD09894970
  • Cas Number : 118778-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98341

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98341
Availability details for
Quantity
  • Availability
  • Add to Cart