ENA358813401 Enamine

1H-1,2,3-triazol-4-ylmethanamine

  • MDL number MFCD09701416
  • Cas Number : 118724-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98429

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-98429
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart