ENA360886649 Enamine

ethyl 2-[3-(3,4-dimethoxyphenyl)-1,2,4-oxadiazol-5-yl]acetate

  • MDL number MFCD21602455
  • Cas Number : 1375184-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93561
Availability details for
Quantity
  • Availability
  • Add to Cart