ENA360886663 Enamine

1-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD21602469
  • Cas Number : 1486485-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95434
Availability details for
Quantity
  • Availability
  • Add to Cart