ENA360886665 Enamine

4,5,6-trimethyl-2H,3H-[1,2]thiazolo[5,4-b]pyridin-3-one

  • MDL number MFCD21602471
  • Cas Number : 1374509-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-95520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95520
Availability details for
Quantity
  • Availability
  • Add to Cart