ENA360886676 Enamine

2-(2-fluorophenyl)azetidine hydrochloride

  • MDL number MFCD21602484
  • Cas Number : 1375231-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95691
Availability details for
Quantity
  • Availability
  • Add to Cart