ENA360886682 Enamine

2-ethanehydrazonoylpyrazine

  • MDL number MFCD21602490
  • Cas Number : 220276-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95949

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95949
Availability details for
Quantity
  • Availability
  • Add to Cart