ENA360886683 Enamine

2-chloro-1-[1-(4-fluorophenyl)-1H-1,2,3-triazol-4-yl]ethan-1-one

  • MDL number MFCD21602491
  • Cas Number : 1375471-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95957
Availability details for
Quantity
  • Availability
  • Add to Cart