ENA360886715 Enamine

2,2,2-trifluoro-1-(4-methyl-1,3-thiazol-2-yl)ethyl 4-methylbenzene-1-sulfonate

  • MDL number MFCD21602530
  • Cas Number : 1375471-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96882
Availability details for
Quantity
  • Availability
  • Add to Cart