ENA360886740 Enamine

4-bromo-2-(bromomethyl)quinoline

  • MDL number MFCD21602559
  • Cas Number : 89446-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97064
Availability details for
Quantity
  • Availability
  • Add to Cart