ENA360886793 Enamine

ethyl 3-amino-3-(2-methylphenyl)propanoate hydrochloride

  • MDL number MFCD21602621
  • Cas Number : 1375471-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97501

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97501
Availability details for
Quantity
  • Availability
  • Add to Cart