ENA360886796 Enamine

N-benzyl-8-(trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-amine

  • MDL number MFCD21602625
  • Cas Number : 1375473-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97551
Availability details for
Quantity
  • Availability
  • Add to Cart