ENA360886823 Enamine

3,5-dimethyl-1,3,4,7-tetraazatricyclo[7.3.0.0,2,6]dodeca-2(6),4-dien-8-one

  • MDL number MFCD21602657
  • Cas Number : 1360059-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97919
Availability details for
Quantity
  • Availability
  • Add to Cart