ENA360886827 Enamine

2-phenyl-4H,5H,6H,7H-furo[2,3-c]pyridine hydrochloride

  • MDL number MFCD21602661
  • Cas Number : 104278-45-7
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-phenyl-4,5,6,7-tetrahydrofuro[2,3-c]pyridine hydrochloride
Inchi Code 1S/C13H13NO.ClH/c1-2-4-10(5-3-1)12-8-11-6-7-14-9-13(11)15-12;/h1-5,8,14H,6-7,9H2;1H
Inchi Key LGIIFZPWSIRNML-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-97939

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-97939
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart