ENA360886840 Enamine

tert-butyl 3-methyl-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-1-carboxylate

  • MDL number MFCD21602677
  • Cas Number : 1375473-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98108
Availability details for
Quantity
  • Availability
  • Add to Cart