ENA360886841 Enamine

5-chloro-2-[2-(dimethylamino)ethoxy]benzoic acid hydrochloride

  • MDL number MFCD21602679
  • Cas Number : 1375473-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98134

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98134
Availability details for
Quantity
  • Availability
  • Add to Cart