ENA360886845 Enamine

5-(methylamino)-2,3-dihydropyridazin-3-one

  • MDL number MFCD24394752
  • Cas Number : 1353498-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98171
Availability details for
Quantity
  • Availability
  • Add to Cart