ENA360886853 Enamine

6-nitro-4-(propylsulfanyl)-1H-indole

  • MDL number MFCD21602693
  • Cas Number : 1375473-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98324
Availability details for
Quantity
  • Availability
  • Add to Cart