ENA360886856 Enamine

{[3-(1-methoxyethyl)-1,2,4-oxadiazol-5-yl]methyl}(methyl)amine

  • MDL number MFCD21602696
  • Cas Number : 1375474-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98475
Availability details for
Quantity
  • Availability
  • Add to Cart