ENA360886865 Enamine

[2,2-dimethyl-3-(methylamino)propyl](methyl)amine dihydrochloride

  • MDL number MFCD21602705
  • Cas Number : 1375474-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98573
Availability details for
Quantity
  • Availability
  • Add to Cart