ENA360886866 Enamine

{[2-(benzyloxy)-3-bromophenyl]methyl}(methyl)amine hydrochloride

  • MDL number MFCD21602706
  • Cas Number : 1375474-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98575
Availability details for
Quantity
  • Availability
  • Add to Cart