ENA360886867 Enamine

1-ethyl-1H-imidazole-5-sulfonyl chloride

  • MDL number MFCD21602707
  • Cas Number : 1375473-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98580
Availability details for
Quantity
  • Availability
  • Add to Cart