ENA360886868 Enamine

2-(methylamino)-2-[4-(trifluoromethyl)phenyl]acetic acid hydrochloride

  • MDL number MFCD21602708
  • Cas Number : 1375471-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98641
Availability details for
Quantity
  • Availability
  • Add to Cart