ENA360886871 Enamine

1-(3-fluoro-4-methoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD21602712
  • Cas Number : 1375472-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98677
Availability details for
Quantity
  • Availability
  • Add to Cart