ENA360886874 Enamine

1-(5-ethyl-1,2,4-oxadiazol-3-yl)cyclopentan-1-amine hydrochloride

  • MDL number MFCD21602715
  • Cas Number : 1375472-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98724
Availability details for
Quantity
  • Availability
  • Add to Cart